SY3H3D681A2E Synthonix Corporation

4-(Pyridin-2-yl)butanoic acid

  • MDL number No MDL found
  • Cas Number : 102879-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50052.html

Safety Information

MSDS https://synthonix.com/50052.html
Availability details for
Quantity
  • Availability
  • Add to Cart