SY3H3D681A2F Synthonix Corporation

2-Isopropyl-4-nitrophenol

  • Cas Number : 60515-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50084.html

Safety Information

MSDS https://synthonix.com/50084.html
Availability details for
Quantity
  • Availability
  • Add to Cart