SY3H3D681A31 Synthonix Corporation

N-(Phenylsulfonyl)glycine Methyl Ester

  • MDL number No MDL found
  • Cas Number : 69398-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/50114.html

Safety Information

MSDS https://synthonix.com/50114.html
Availability details for
Quantity
  • Availability
  • Add to Cart