SY3H3D681A35 Synthonix Corporation

2-(Pyridin-4-yl)morpholine

  • MDL number No MDL found
  • Cas Number : 1018656-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50172.html

Safety Information

MSDS https://synthonix.com/50172.html
Availability details for
Quantity
  • Availability
  • Add to Cart