SY3H3D681A38 Synthonix Corporation

2-(Piperidin-1-yl)thiazole-4-carboxylic acid

  • Cas Number : 952182-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50192.html

Safety Information

MSDS https://synthonix.com/50192.html
Availability details for
Quantity
  • Availability
  • Add to Cart