SY3H3D681A3B Synthonix Corporation

4-Propylbenzene-1,3-diol

  • MDL number No MDL found
  • Cas Number : 18979-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50281.html

Safety Information

MSDS https://synthonix.com/50281.html
Availability details for
Quantity
  • Availability
  • Add to Cart