SY3H3D681A3C Synthonix Corporation

Propyl benzenesulfonate

  • Cas Number : 80-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50300.html

Safety Information

MSDS https://synthonix.com/50300.html
Availability details for
Quantity
  • Availability
  • Add to Cart