SY3H3D681A3D Synthonix Corporation

Isopropyl benzenesulfonate

  • MDL number No MDL found
  • Cas Number : 6214-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/50301.html

Safety Information

MSDS https://synthonix.com/50301.html
Availability details for
Quantity
  • Availability
  • Add to Cart