SY3H3D681A3E Synthonix Corporation

Phenethyl methanesulfonate

  • MDL number No MDL found
  • Cas Number : 20020-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/50302.html

Safety Information

MSDS https://synthonix.com/50302.html
Availability details for
Quantity
  • Availability
  • Add to Cart