SY3H3D681A3F Synthonix Corporation

3-Isopropylaniline

  • MDL number No MDL found
  • Cas Number : 5369-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/50307.html

Safety Information

MSDS https://synthonix.com/50307.html
Availability details for
Quantity
  • Availability
  • Add to Cart