SY3H3D681A41 Synthonix Corporation

4-(Pyrrolidin-1-yl)pyridin-2-amine

  • Cas Number : 722550-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50323.html

Safety Information

MSDS https://synthonix.com/50323.html
Availability details for
Quantity
  • Availability
  • Add to Cart