SY3H3D681A92 Synthonix Corporation

4-(Pyridin-2-yl)butan-1-ol

  • MDL number No MDL found
  • Cas Number : 17945-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50340.html

Safety Information

MSDS https://synthonix.com/50340.html
Availability details for
Quantity
  • Availability
  • Add to Cart