SY3H3D681A98 Synthonix Corporation

6-(Piperidin-1-yl)pyrimidin-4-amine

  • MDL number No MDL found
  • Cas Number : 69206-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/50425.html

Safety Information

MSDS https://synthonix.com/50425.html
Availability details for
Quantity
  • Availability
  • Add to Cart