SY3H3D681A99 Synthonix Corporation

N-(Piperidin-4-yl)pyrimidin-2-amine

  • MDL number No MDL found
  • Cas Number : 69385-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50426.html

Safety Information

MSDS https://synthonix.com/50426.html
Availability details for
Quantity
  • Availability
  • Add to Cart