SY3H3D681AA0 Synthonix Corporation

(R)-1-(Pyrrolidin-3-yl)piperidine

  • Cas Number : 913812-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50567.html

Safety Information

MSDS https://synthonix.com/50567.html
Availability details for
Quantity
  • Availability
  • Add to Cart