SY3H3D681AA6 Synthonix Corporation

6-(1H-Pyrazol-1-yl)nicotinic acid

  • MDL number No MDL found
  • Cas Number : 253315-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51143.html

Safety Information

MSDS https://synthonix.com/51143.html
Availability details for
Quantity
  • Availability
  • Add to Cart