SY3H3D681AA8 Synthonix Corporation

4-Phenyloxazole

  • Cas Number : 20662-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51158.html

Safety Information

MSDS https://synthonix.com/51158.html
Availability details for
Quantity
  • Availability
  • Add to Cart