SY3H3D681AA9 Synthonix Corporation

3-(Phenylsulfonyl)acrylonitrile

  • MDL number No MDL found
  • Cas Number : 1424-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/51192.html

Safety Information

MSDS https://synthonix.com/51192.html
Availability details for
Quantity
  • Availability
  • Add to Cart