SY3H3D681AB2 Synthonix Corporation

1-Phenyl-1H-pyrazol-5-ol

  • MDL number No MDL found
  • Cas Number : 876-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51348.html

Safety Information

MSDS https://synthonix.com/51348.html
Availability details for
Quantity
  • Availability
  • Add to Cart