SY3H3D681AD4 Synthonix Corporation

1-(Pyridin-4-yl)cyclopropanecarboxylic acid

  • MDL number MFCD09864787
  • Cas Number : 920297-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51790.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51790.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart