SY3H3D681AE6 Synthonix Corporation

N-(Piperidin-3-yl)pyrimidin-2-amine hydrochloride

  • Cas Number : 1185312-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52190.html

Safety Information

MSDS https://synthonix.com/52190.html
Availability details for
Quantity
  • Availability
  • Add to Cart