SY3H3D681AE8 Synthonix Corporation

N-(Piperidin-4-yl)pyrimidin-2-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 950649-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52191.html

Safety Information

MSDS https://synthonix.com/52191.html
Availability details for
Quantity
  • Availability
  • Add to Cart