SY3H3D681AEA Synthonix Corporation

1-(Piperidin-4-yl)pyrrolidin-2-one hydrochloride

  • MDL number No MDL found
  • Cas Number : 548769-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52198.html

Safety Information

MSDS https://synthonix.com/52198.html
Availability details for
Quantity
  • Availability
  • Add to Cart