SY3H3D681AF0 Synthonix Corporation

1-(Pyridin-4-yl)butan-1-amine dihydrochloride

  • MDL number No MDL found
  • Cas Number : 2097938-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52255.html

Safety Information

MSDS https://synthonix.com/52255.html
Availability details for
Quantity
  • Availability
  • Add to Cart