SY3H3D681AF8 Synthonix Corporation

1-(Piperidin-4-yl)piperazine trihydrochloride

  • MDL number No MDL found
  • Cas Number : 1217074-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52262.html

Safety Information

MSDS https://synthonix.com/52262.html
Availability details for
Quantity
  • Availability
  • Add to Cart