SY3H3D681B06 Synthonix Corporation

((Prop-2-yn-1-yloxy)methyl)benzene

  • MDL number No MDL found
  • Cas Number : 4039-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52414.html

Safety Information

MSDS https://synthonix.com/52414.html
Availability details for
Quantity
  • Availability
  • Add to Cart