SY3H3D681B08 Synthonix Corporation

2-Phenyl-1,3-dioxan-5-one

  • MDL number No MDL found
  • Cas Number : 52941-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52455.html

Safety Information

MSDS https://synthonix.com/52455.html
Availability details for
Quantity
  • Availability
  • Add to Cart