SY3H3D681C12 Synthonix Corporation

N-(1-Phenylvinyl)acetamide

  • MDL number No MDL found
  • Cas Number : 57957-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52545.html

Safety Information

MSDS https://synthonix.com/52545.html
Availability details for
Quantity
  • Availability
  • Add to Cart