SY3H3D681C16 Synthonix Corporation

1-Pyridin-4-ylpiperidin-4-one

  • Cas Number : 126832-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52686.html

Safety Information

MSDS https://synthonix.com/52686.html
Availability details for
Quantity
  • Availability
  • Add to Cart