SY3H3D681C18 Synthonix Corporation

3-Propyl-1H-pyrrolo[2,3-c]pyridin-7(6H)-one

  • MDL number No MDL found
  • Cas Number : 1493855-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/52690.html

Safety Information

MSDS https://synthonix.com/52690.html
Availability details for
Quantity
  • Availability
  • Add to Cart