SY3H3D681C19 Synthonix Corporation

N,N'-(1,2-Phenylene)diacetamide

  • MDL number No MDL found
  • Cas Number : 2050-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52695.html

Safety Information

MSDS https://synthonix.com/52695.html
Availability details for
Quantity
  • Availability
  • Add to Cart