SY3H3D681C1E Synthonix Corporation

4-Phenyltetrahydrofuran-3-ol

  • Cas Number : 1420794-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/52796.html

Safety Information

MSDS https://synthonix.com/52796.html
Availability details for
Quantity
  • Availability
  • Add to Cart