SY3H3D681C20 Synthonix Corporation

1-Phenylpyrrolidine

  • MDL number No MDL found
  • Cas Number : 4096-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52876.html

Safety Information

MSDS https://synthonix.com/52876.html
Availability details for
Quantity
  • Availability
  • Add to Cart