SY3H3D681C23 Synthonix Corporation

1-(5-Isopropylpyridin-2-yl)ethanone

  • MDL number No MDL found
  • Cas Number : 137853-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52923.html

Safety Information

MSDS https://synthonix.com/52923.html
Availability details for
Quantity
  • Availability
  • Add to Cart