SY3H3D681C27 Synthonix Corporation

1-(2-Isopropoxypyridin-3-yl)ethanone

  • MDL number No MDL found
  • Cas Number : 1551553-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52960.html

Safety Information

MSDS https://synthonix.com/52960.html
Availability details for
Quantity
  • Availability
  • Add to Cart