SY3H3D681C28 Synthonix Corporation

(R)-2-((1-Phenylethyl)amino)acetic acid

  • Cas Number : 78397-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52966.html

Safety Information

MSDS https://synthonix.com/52966.html
Availability details for
Quantity
  • Availability
  • Add to Cart