SY3H3D681C2A Synthonix Corporation

2-(Piperidin-3-yl)pyridine

  • Cas Number : 40864-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53025.html

Safety Information

MSDS https://synthonix.com/53025.html
Availability details for
Quantity
  • Availability
  • Add to Cart