SY3H3D681C31 Synthonix Corporation

Isopropyl 3,4-diaminobenzoate

  • MDL number No MDL found
  • Cas Number : 121649-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53047.html

Safety Information

MSDS https://synthonix.com/53047.html
Availability details for
Quantity
  • Availability
  • Add to Cart