SY3H3D681C32 Synthonix Corporation

2-Propyl-1H-imidazole-4,5-dicarboxylic acid dimethyl ester

  • MDL number No MDL found
  • Cas Number : 124750-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/53056.html

Safety Information

MSDS https://synthonix.com/53056.html
Availability details for
Quantity
  • Availability
  • Add to Cart