SY3H3D681C38 Synthonix Corporation

3-Pentylpyridine

  • MDL number No MDL found
  • Cas Number : 1802-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53110.html

Safety Information

MSDS https://synthonix.com/53110.html
Availability details for
Quantity
  • Availability
  • Add to Cart