SY3H3D681C3A Synthonix Corporation

1-(Pyridin-3-yl)-1,4-diazepane

  • MDL number No MDL found
  • Cas Number : 223796-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53123.html

Safety Information

MSDS https://synthonix.com/53123.html
Availability details for
Quantity
  • Availability
  • Add to Cart