SY3H3D681C45 Synthonix Corporation

1,4,7,10,13-Pentaazacyclopentadecane

  • Cas Number : 295-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53370.html

Safety Information

MSDS https://synthonix.com/53370.html
Availability details for
Quantity
  • Availability
  • Add to Cart