SY3H3D681C46 Synthonix Corporation

2,2'-(Perchloro-1,2-phenylene)diacetonitrile

  • MDL number No MDL found
  • Cas Number : 60069-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53374.html

Safety Information

MSDS https://synthonix.com/53374.html
Availability details for
Quantity
  • Availability
  • Add to Cart