SY3H3D681C49 Synthonix Corporation

1-Phenyl-3-(trifluoromethyl)-1H-pyrazole

  • MDL number No MDL found
  • Cas Number : 99498-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/53523.html

Safety Information

MSDS https://synthonix.com/53523.html
Availability details for
Quantity
  • Availability
  • Add to Cart