SY3H3D681C4A Synthonix Corporation

2-Phenyl-4-(trifluoromethyl)-1H-imidazole

  • Cas Number : 33469-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53524.html

Safety Information

MSDS https://synthonix.com/53524.html
Availability details for
Quantity
  • Availability
  • Add to Cart