SY3H3D681C4E Synthonix Corporation

3H-Pyrimido[5,4-b]indol-4(5H)-one

  • MDL number MFCD00141061
  • Cas Number : 61553-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53545.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53545.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart