SY3H3D681C62 Synthonix Corporation

1-Phenyl-1H-imidazole-5-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 135417-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53665.html

Safety Information

MSDS https://synthonix.com/53665.html
Availability details for
Quantity
  • Availability
  • Add to Cart