SY3H3D681C63 Synthonix Corporation

5-Phenyl-1H-imidazole-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 41270-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/53666.html

Safety Information

MSDS https://synthonix.com/53666.html
Availability details for
Quantity
  • Availability
  • Add to Cart