SY3H3D681C65 Synthonix Corporation

4-(Prop-1-yn-1-yl)benzoic acid

  • MDL number No MDL found
  • Cas Number : 223562-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53794.html

Safety Information

MSDS https://synthonix.com/53794.html
Availability details for
Quantity
  • Availability
  • Add to Cart