SY3H3D681C67 Synthonix Corporation

1-Phenyl-1-cyclopropanecarbonitrile

  • MDL number No MDL found
  • Cas Number : 935-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/53837.html

Safety Information

MSDS https://synthonix.com/53837.html
Availability details for
Quantity
  • Availability
  • Add to Cart